siRNA Details
| Gene Symbol: LOC101929705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125864.1 | 5 | 1130 | |||
| NR_125865.1 | 4 | 914 | ||||
| GenBank mRNA | AK024736.1 | - | 5 | 1130 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929705| Gene Symbol: LOC101929705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125864.1 | 5 | 1130 | |||
| NR_125865.1 | 4 | 914 | ||||
| GenBank mRNA | AK024736.1 | - | 5 | 1130 | ||