siRNA Details
Gene Symbol: LOC101929705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125864.1 | 5 | 2190 | |||
NR_125865.1 | 4 | 1974 | ||||
GenBank mRNA | AK024736.1 | - | 5 | 2184 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000422059.5 | - | 5 | 2190 | Chr. 6 - 43995723 - 44074652 [-] on Build GRCh38 |
