siRNA Details
| Gene Symbol: LOC284628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094692.1 | - | 2 | 299 | ||
| DA148243.1 | - | 3 | 265 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA164101.1 | - | 2 | 299 | Chr. 1 - 1600439 - 1607180 [+] on Build GRCh38 | |

LOC284628
Genomic Map