siRNA Details
| Gene Symbol: SPRY4-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131221.1 | 1 | 291 | |||
| GenBank mRNA | AK024556.1 | - | 1 | 292 | ||
| Gene Symbol: SPRY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017009910.1 | XP_016865399.1 | 1 | 285 | ||

SPRY4-IT1
Genomic Map