siRNA Details
| Gene Symbol: SPRY4-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024556.1 | - | 1 | 482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRY4-IT1| Gene Symbol: SPRY4-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024556.1 | - | 1 | 482 | ||