siRNA Details
| Gene Symbol: LINC01153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037739.1 | - | 2 | 1678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01153| Gene Symbol: LINC01153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037739.1 | - | 2 | 1678 | ||