siRNA Details
| Gene Symbol: LINC01153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037739.1 | - | 2 | 2691 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429809.1 | - | 2 | 2686 | Chr. 10 - 121178700 - 121185966 [+] on Build GRCh38 | |

LINC01153
Genomic Map