siRNA Details
| Gene Symbol: LPEQ6126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY358194.1 | - | 1 | 883 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPEQ6126| Gene Symbol: LPEQ6126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY358194.1 | - | 1 | 883 | ||