siRNA Details
| Gene Symbol: LOC101928126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134561.1 | 2 | 182 | |||
| GenBank mRNA | AK124146.1 | - | 1 | 1503 | ||
| BG771030.1 | - | 2 | 183 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF887353.1 | - | 1 | 427 | Chr. 21 - 33976305 - 33977039 [+] on Build GRCh38 | |

LOC101928126
Genomic Map