siRNA Details
| Gene Symbol: LINC00315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF427488.1 | - | 3 | 2191 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BP378314.1 | - | 1 | 294 | Chr. 21 - 45300102 - 45300669 [-] on Build GRCh38 | |

LINC00315
Genomic Map