siRNA Details
| Gene Symbol: LINC00358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046995.1 | 1 | 36 | |||
| GenBank mRNA | BC037867.2 | - | 1 | 36 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI464936.1 | - | 1 | 41 | Chr. 13 - 62004093 - 62029548 [-] on Build GRCh38 | |

LINC00358
Genomic Map