siRNA Details
| Gene Symbol: ZNF224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC002889.1 | - | 1 | 649 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY800134.1 | - | 1 | 593 | Chr. 19 - 44094443 - 44095046 [+] on Build GRCh38 | |

ZNF224
Genomic Map