siRNA Details
| Gene Symbol: CARMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126541.1 | - | 1 | 2629 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF995194.1 | - | 1 | 379 | Chr. 5 - 149409520 - 149410269 [+] on Build GRCh38 | |

CARMN
Genomic Map