siRNA Details
| Gene Symbol: TNRC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025277.1 | - | 2138 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA842111.1 | - | 1 | 181 | Chr. 16 - 24818623 - 24819013 [+] on Build GRCh38 | |

TNRC6A
Genomic Map