siRNA Details
| Gene Symbol: C6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006715372.2 | XP_006715435.1 | 1 | 1428 | ||
| GenBank mRNA | AK057638.1 | - | 1 | 1428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6orf163| Gene Symbol: C6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006715372.2 | XP_006715435.1 | 1 | 1428 | ||
| GenBank mRNA | AK057638.1 | - | 1 | 1428 | ||