siRNA Details
| Gene Symbol: TRIM52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB209243.1 | - | 1 | 1899 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC026298.1 | - | 1 | 366 | Chr. 4 - 186116746 - 186120499 [+] on Build GRCh38 | |

TRIM52
Genomic Map