siRNA Details
| Gene Symbol: RNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134337.2 | NP_001127809.1 | 6 | 6592 | ||
| NM_001134338.2 | NP_001127810.1 | 7 | 6661 | |||
| NM_001321749.1 | NP_001308678.1 | 6 | 6828 | |||
| NM_007219.4 | NP_009150.1 | 6 | 6884 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR613361.1 | - | 1 | 627 | Chr. 20 - 3927354 - 3928706 [-] on Build GRCh38 | |

RNF24
Genomic Map