siRNA Details
| Gene Symbol: RNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134337.2 | NP_001127809.1 | 6 | 7028 | ||
| NM_001134338.2 | NP_001127810.1 | 7 | 7097 | |||
| NM_001321749.1 | NP_001308678.1 | 6 | 7264 | |||
| NM_007219.4 | NP_009150.1 | 6 | 7320 | |||

RNF24
Genomic Map