siRNA Details
| Gene Symbol: LOC101929268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105002.1 | 4 | 1748 | |||
| GenBank mRNA | AK092055.1 | - | 4 | 1748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929268| Gene Symbol: LOC101929268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105002.1 | 4 | 1748 | |||
| GenBank mRNA | AK092055.1 | - | 4 | 1748 | ||