siRNA Details
| Gene Symbol: NBPF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX538005.1 | - | 2 | 291 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF370406.1 | - | 2045 | Chr. - - [-] on Build | ||

NBPF10
Genomic Map