siRNA Details
| Gene Symbol: ATP6V1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127836.1 | - | 1 | 814 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF370403.1 | - | 1 | 830 | Chr. 2 - 10776985 - 10779707 [+] on Build GRCh38 | |

ATP6V1C2
Genomic Map