siRNA Details
| Gene Symbol: LOC100505915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125434.1 | 4 | 2570 | |||
| GenBank mRNA | AK125313.1 | - | 4 | 2570 | ||
| BC017937.1 | - | 1 | 1298 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569858.1 | - | 4 | 2570 | Chr. 16 - 15094411 - 15109197 [+] on Build GRCh38 | |

LOC100505915
Genomic Map