siRNA Details
| Gene Symbol: KCTD21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124949.1 | - | 2 | 187 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD21-AS1| Gene Symbol: KCTD21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124949.1 | - | 2 | 187 | ||