siRNA Details
| Gene Symbol: KCTD21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102280.1 | 2 | 344 | |||
| GenBank mRNA | AK124949.1 | - | 2 | 359 | ||
| DA737651.1 | - | 2 | 295 | |||
| DB082989.1 | - | 2 | 343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD21-AS1| Gene Symbol: KCTD21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102280.1 | 2 | 344 | |||
| GenBank mRNA | AK124949.1 | - | 2 | 359 | ||
| DA737651.1 | - | 2 | 295 | |||
| DB082989.1 | - | 2 | 343 | |||