siRNA Details
| Gene Symbol: LOC645485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135046.1 | 2 | 241 | |||
| GenBank mRNA | BC039526.1 | - | 2 | 241 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM563950.1 | - | 2 | 261 | Chr. 12 - 30755074 - 30780230 [+] on Build GRCh38 | |

LOC645485
Genomic Map