siRNA Details
| Gene Symbol: LOC645485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135046.1 | 2 | 1470 | |||
| GenBank mRNA | BC039526.1 | - | 2 | 1470 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC645485| Gene Symbol: LOC645485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135046.1 | 2 | 1470 | |||
| GenBank mRNA | BC039526.1 | - | 2 | 1470 | ||