siRNA Details
| Gene Symbol: SPPL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005254722.3 | XP_005254779.1 | 16 | 4903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPPL2A| Gene Symbol: SPPL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005254722.3 | XP_005254779.1 | 16 | 4903 | ||