siRNA Details
| Gene Symbol: SPPL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005254722.3 | XP_005254779.1 | 16 | 3738 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC036610.1 | - | 1 | 259 | Chr. 15 - 50704531 - 50706116 [-] on Build GRCh38 | |

SPPL2A
Genomic Map