siRNA Details
| Gene Symbol: TOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC020727.1 | - | 3 | 457 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK022060.1 | - | 2 | 2244 | Chr. 14 - 21483483 - 21488067 [+] on Build GRCh38 | |

TOX4
Genomic Map