siRNA Details
| Gene Symbol: RAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055170.1 | - | 1 | 1560 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI910311.1 | - | 1 | 134 | Chr. 20 - 57377218 - 57377989 [+] on Build GRCh38 | |

RAE1
Genomic Map