siRNA Details
| Gene Symbol: STAM-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110370.1 | 1 | 1724 | |||
| GenBank mRNA | AK123677.1 | - | 1 | 1685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STAM-AS1| Gene Symbol: STAM-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110370.1 | 1 | 1724 | |||
| GenBank mRNA | AK123677.1 | - | 1 | 1685 | ||