siRNA Details
| Gene Symbol: LOC729652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135198.1 | 1 | 2098 | |||
| GenBank mRNA | AL834536.1 | - | 1 | 2098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC729652| Gene Symbol: LOC729652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135198.1 | 1 | 2098 | |||
| GenBank mRNA | AL834536.1 | - | 1 | 2098 | ||