siRNA Details
| Gene Symbol: NCOA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012970.1 | XP_016868459.1 | 1 | 4709 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK025045.1 | - | 1 | 265 | Chr. 8 - 70218200 - 70220303 [-] on Build GRCh38 | |

NCOA2
Genomic Map