siRNA Details
| Gene Symbol: KIAA1656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046312.1 | 2 | 1978 | |||
| GenBank mRNA | AB051443.1 | - | 1 | 1343 | ||
| CR456347.1 | - | 1 | 441 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1656| Gene Symbol: KIAA1656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046312.1 | 2 | 1978 | |||
| GenBank mRNA | AB051443.1 | - | 1 | 1343 | ||
| CR456347.1 | - | 1 | 441 | |||