siRNA Details
Gene Symbol: KIAA1656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046312.1 | 2 | 1689 | |||
GenBank mRNA | AB051443.1 | - | 1 | 1054 | ||
CR456347.1 | - | 1 | 152 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIAA1656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046312.1 | 2 | 1689 | |||
GenBank mRNA | AB051443.1 | - | 1 | 1054 | ||
CR456347.1 | - | 1 | 152 |