siRNA Details
| Gene Symbol: LOC284570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC033722.1 | - | 1 | 1222 | ||
| BC040156.1 | - | 1 | 1330 | |||
| Gene Symbol: GPR37L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510158.2 | XP_011508460.1 | 2 | 3830 | ||

LOC284570
Genomic Map