siRNA Details
| Gene Symbol: LOC284570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040156.1 | - | 1 | 528 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG910726.1 | - | 1 | 527 | Chr. 1 - 202130056 - 202130905 [+] on Build GRCh38 | |

LOC284570
Genomic Map