siRNA Details
| Gene Symbol: CAMTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242701.1 | NP_001229630.1 | 4 | 965 | ||
| NR_038934.1 | 4 | 979 | ||||
| GenBank mRNA | AF086286.1 | - | 1 | 376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAMTA1| Gene Symbol: CAMTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242701.1 | NP_001229630.1 | 4 | 965 | ||
| NR_038934.1 | 4 | 979 | ||||
| GenBank mRNA | AF086286.1 | - | 1 | 376 | ||