siRNA Details
| Gene Symbol: LOC100131840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057355.1 | - | 1 | 1198 | ||
| Gene Symbol: CCDC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011533673.2 | XP_011531975.1 | 1 | 361 | ||

LOC100131840
Genomic Map