siRNA Details
| Gene Symbol: LCORL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153686.8 | NP_710153.2 | 7 | 3860 | ||
| NR_136669.1 | 5 | 3686 | ||||
| XM_011513821.2 | XP_011512123.1 | 9 | 8591 | |||
| XM_011513822.2 | XP_011512124.1 | 8 | 8564 | |||
| XM_011513823.2 | XP_011512125.1 | 8 | 8437 | |||
| XM_011513826.2 | XP_011512128.1 | 6 | 3644 | |||
| XM_017007961.1 | XP_016863450.1 | 8 | 9873 | |||
| XM_017007962.1 | XP_016863451.1 | 8 | 8515 | |||
| XM_017007963.1 | XP_016863452.1 | 8 | 8818 | |||
| GenBank mRNA | AL133031.1 | - | 1 | 1989 | ||

LCORL
Genomic Map