siRNA Details
| Gene Symbol: TTLL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005271208.2 | XP_005271265.1 | 20 | 5696 | ||
| XM_017002347.1 | XP_016857836.1 | 22 | 5905 | |||
| XM_017002349.1 | XP_016857838.1 | 20 | 5660 | |||
| GenBank mRNA | BC042023.1 | - | 1 | 1018 | ||

TTLL7
Genomic Map