siRNA Details
| Gene Symbol: KAZN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011541074.2 | XP_011539376.1 | 16 | 3503 | ||
| XM_011541075.2 | XP_011539377.1 | 14 | 3159 | |||
| XM_011541076.2 | XP_011539378.1 | 14 | 3304 | |||
| XM_011541077.2 | XP_011539379.1 | 14 | 4283 | |||
| XM_011541081.2 | XP_011539383.1 | 8 | 1983 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK095631.1 | - | 2 | 3123 | Chr. 1 - 15109946 - 15113444 [+] on Build GRCh38 | |

KAZN
Genomic Map