siRNA Details
| Gene Symbol: SRSF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021742.1 | - | 1 | 880 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX951495.1 | - | 1 | 201 | Chr. 1 - 70227686 - 70228377 [+] on Build GRCh38 | |

SRSF11
Genomic Map