siRNA Details
| Gene Symbol: LOC101929526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109955.1 | 1 | 161 | |||
| GenBank mRNA | BC042559.1 | - | 1 | 161 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929526| Gene Symbol: LOC101929526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109955.1 | 1 | 161 | |||
| GenBank mRNA | BC042559.1 | - | 1 | 161 | ||