siRNA Details
| Gene Symbol: OPCML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006718846.2 | XP_006718909.1 | 8 | 9880 | ||
| XM_011542856.2 | XP_011541158.1 | 9 | 10193 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK054706.1 | - | 2 | 2267 | Chr. 11 - 132411563 - 132414196 [-] on Build GRCh38 | |

OPCML
Genomic Map