siRNA Details
| Gene Symbol: TCEB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW451236.1 | - | 1 | 231 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124839.1 | - | 1 | 999 | Chr. 1 - 23760519 - 23762843 [-] on Build GRCh38 | |

TCEB3
Genomic Map