siRNA Details
| Gene Symbol: RAB11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206836.1 | NP_001193765.1 | 5 | 4680 | ||
| NM_004663.4 | NP_004654.1 | 5 | 4775 | |||
| GenBank mRNA | BC047114.1 | - | 1 | 1067 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG621535.1 | - | 1 | 553 | Chr. 15 - 65891285 - 65891976 [+] on Build GRCh38 | |

RAB11A
Genomic Map