siRNA Details
| Gene Symbol: FAM177A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040538.1 | - | 1973 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM177A1| Gene Symbol: FAM177A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040538.1 | - | 1973 | |||