siRNA Details
| Gene Symbol: ZNF81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005272600.3 | XP_005272657.1 | 5 | 10601 | ||
| XM_011543899.2 | XP_011542201.1 | 5 | 12207 | |||
| XM_011543900.2 | XP_011542202.1 | 6 | 10662 | |||
| XM_017029486.1 | XP_016884975.1 | 6 | 10720 | |||
| XM_017029487.1 | XP_016884976.1 | 5 | 10953 | |||
| GenBank mRNA | AK026716.1 | - | 1 | 1222 | ||

ZNF81
Genomic Map