siRNA Details
| Gene Symbol: ZNF81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005272600.3 | XP_005272657.1 | 5 | 10200 | ||
| XM_011543899.2 | XP_011542201.1 | 5 | 11806 | |||
| XM_011543900.2 | XP_011542202.1 | 6 | 10261 | |||
| XM_017029486.1 | XP_016884975.1 | 6 | 10319 | |||
| XM_017029487.1 | XP_016884976.1 | 5 | 10552 | |||
| GenBank mRNA | AK026716.1 | - | 1 | 821 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA571914.1 | - | 1 | 599 | Chr. X - 47923941 - 47924804 [+] on Build GRCh38 | |

ZNF81
Genomic Map